missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vancomycin hydrochloride, Molecular Biology Grade

Product Code. p-7042509
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15495089

Brand: Thermo Scientific Chemicals J62790.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vancomycin hydrochloride (vancomycin HCl, CAS # 1404-93-9) is the hydrochloride salt of the bactericidal agent, vancomycin. | CAS: 1404-93-9 | C66H76Cl3N9O24 | 1485.72 g/mol

Inhibits bacterial mucopeptide synthesis.Vancomycin hydrochloride is used as an antiviral and anti bacterial. binds to bacterial cell wall. It exhibits potent activity against Gram positive bacteria and is highly effective against MRSA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Vancomycin hydrochloride is a glycopeptide antibiotic that prevents cell wall synthesis
  • Originally obtained from the bacterial strain Streptomyces Orientalis
  • Vancomycin binds to D-alanyl-D-alanine motifs in cell wall precursors and leads to cell death
  • It is active against some penicillin-resistant bacteria

Application

  • Vancomycin hydrochloride is intended for use in the research of infections and for antibiotic resistance testing
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1404-93-9
Molecular Formula C66H76Cl3N9O24
Molecular Weight (g/mol) 1485.72
MDL Number MFCD03613611,MFCD03613611
InChI Key LCTORFDMHNKUSG-UHFFFAOYNA-N
PubChem CID 124080918
IUPAC Name 48-({3-[(4-amino-5-hydroxy-4,6-dimethyloxan-2-yl)oxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl}oxy)-22-(carbamoylmethyl)-5,15-dichloro-2,18,32,35,37-pentahydroxy-19-[4-methyl-2-(methylamino)pentanamido]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentaazaoctacyclo[26.14.2.2³,⁶.2¹⁴,¹⁷.1⁸,¹².1²⁹,³³.0¹⁰,²⁵.0³⁴,³⁹]pentaconta-3,5,8(48),9,11,14,16,29(45),30,32,34,36,38,46,49-pentadecaene-40-carboxylic acid hydrochloride
SMILES Cl.CNC(CC(C)C)C(=O)NC1C(O)C2=CC(Cl)=C(OC3=CC4=CC(OC5=C(Cl)C=C(C=C5)C(O)C5NC(=O)C(NC(=O)C4NC(=O)C(CC(N)=O)NC1=O)C1=CC(=C(O)C=C1)C1=C(O)C=C(O)C=C1C(NC5=O)C(O)=O)=C3OC1OC(CO)C(O)C(O)C1OC1CC(C)(N)C(O)C(C)O1)C=C2

Specifications

Type Vancomycin Hydrochloride
Physical Form Powder
Color White
Quantity 5 g
Sensitivity Hygroscopic
Beilstein 3704657
Merck Index 149929
Solubility Information Soluble in water; Slightly soluble in methanol,ethanol and dimethylsulfoxide.
Formula Weight 1485.73
Grade Molecular Biology
Chemical Name or Material Vancomycin hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Vancomycin hydrochloride, Molecular Biology Grade
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Vancomycin hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.