missing translation for 'onlineSavingsMsg'
Learn More

Vancomycin hydrochloride, MedChemExpress

Product Code. p-200099062
Encompass_Preferred
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 16499807

Brand: Medchem Express HY17362

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vancomycin hydrochloride is an antibiotic for the treatment of bacterial infections. It acts by inhibiting the second stage of cell wall synthesis of susceptible bacteria. Vancomycin also alters the permeability of the cell membrane and selectively inhibits ribonucleic acid synthesis.

Vancomycin hydrochloride is an antibiotic for the treatment of bacterial infections. It acts by inhibiting the second stage of cell wall synthesis of susceptible bacteria. Vancomycin also alters the permeability of the cell membrane and selectively inhibits ribonucleic acid synthesis.

Specifications

Chemical Name or Material Vancomycin hydrochloride
Physical Form Solid
CAS 1404-93-9
Color Light Nude
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C66H76Cl3N9O24
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 33.33 mg/mL (22.43 mM; Need ultrasonic) ∣DMSO : 24 mg/mL (16.15 mM; Need ultrasonic and warming)
SMILES ClC1=CC([C@H]([C@@H](NC([C@H](NC)CC(C)C)=O)C(N[C@@H](CC(N)=O)C(N[C@@]2([H])C(N[C@]3([H])C4=CC(C5=C(C=C(O)C=C5O)[C@](C(O)=O)([H])NC([C@](NC3=O)([H])[C@H](O)C6=CC=C7C(Cl)=C6)=O)=C(O)C=C4)=O)=O)=O)O)=CC=C1OC8=C(C(O7)=CC2=C8)O[C@H]9[C@@H]([C@H]([C@H](O)[C@@H](CO)O9)O)O[C@@]%10([H])C[C@](N)([C@H](O)[C@H](C)O%10)C.Cl
Molecular Weight (g/mol) 1485.71
Formula Weight 1485.71
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.