missing translation for 'onlineSavingsMsg'
Learn More

Vancomycin hydrochloride, MedChemExpress

Product Code. 16499807
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499807 5 g 5g
16469865 250 mg 250mg
16431669 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16499807 Supplier Medchem Express Supplier No. HY17362
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Vancomycin hydrochloride is an antibiotic for the treatment of bacterial infections. It acts by inhibiting the second stage of cell wall synthesis of susceptible bacteria. Vancomycin also alters the permeability of the cell membrane and selectively inhibits ribonucleic acid synthesis.

Vancomycin hydrochloride is an antibiotic for the treatment of bacterial infections. It acts by inhibiting the second stage of cell wall synthesis of susceptible bacteria. Vancomycin also alters the permeability of the cell membrane and selectively inhibits ribonucleic acid synthesis.

Specifications

Chemical Name or Material Vancomycin hydrochloride
Physical Form Solid
CAS 1404-93-9
Color Light Nude
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C66H76Cl3N9O24
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 33.33 mg/mL (22.43 mM; Need ultrasonic) ∣DMSO : 24 mg/mL (16.15 mM; Need ultrasonic and warming)
SMILES ClC1=CC([C@H]([C@@H](NC([C@H](NC)CC(C)C)=O)C(N[C@@H](CC(N)=O)C(N[C@@]2([H])C(N[C@]3([H])C4=CC(C5=C(C=C(O)C=C5O)[C@](C(O)=O)([H])NC([C@](NC3=O)([H])[C@H](O)C6=CC=C7C(Cl)=C6)=O)=C(O)C=C4)=O)=O)=O)O)=CC=C1OC8=C(C(O7)=CC2=C8)O[C@H]9[C@@H]([C@H]([C@H](O)[C@@H](CO)O9)O)O[C@@]%10([H])C[C@](N)([C@H](O)[C@H](C)O%10)C.Cl
Molecular Weight (g/mol) 1485.71
Formula Weight 1485.71
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.