missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vancomycin hydrochloride, MedChemExpress
Description
Vancomycin hydrochloride is an antibiotic for the treatment of bacterial infections. It acts by inhibiting the second stage of cell wall synthesis of susceptible bacteria. Vancomycin also alters the permeability of the cell membrane and selectively inhibits ribonucleic acid synthesis.
Specifications
Specifications
| Chemical Name or Material | Vancomycin hydrochloride |
| Physical Form | Solid |
| CAS | 1404-93-9 |
| Color | Light Nude |
| Quantity | 1 g |
| Purity Grade Notes | Research |
| Molecular Formula | C66H76Cl3N9O24 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | H2O : 33.33 mg/mL (22.43 mM; Need ultrasonic) ∣DMSO : 24 mg/mL (16.15 mM; Need ultrasonic and warming) |
| SMILES | ClC1=CC([C@H]([C@@H](NC([C@H](NC)CC(C)C)=O)C(N[C@@H](CC(N)=O)C(N[C@@]2([H])C(N[C@]3([H])C4=CC(C5=C(C=C(O)C=C5O)[C@](C(O)=O)([H])NC([C@](NC3=O)([H])[C@H](O)C6=CC=C7C(Cl)=C6)=O)=C(O)C=C4)=O)=O)=O)O)=CC=C1OC8=C(C(O7)=CC2=C8)O[C@H]9[C@@H]([C@H]([C@H](O)[C@@H](CO)O9)O)O[C@@]%10([H])C[C@](N)([C@H](O)[C@H](C)O%10)C.Cl |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?