missing translation for 'onlineSavingsMsg'
Learn More

Vancomycin Hydrochloride, Fisher BioReagents

Product Code. 10014257
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
missing translation for 'unitSize':
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
10014257 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10014257 missing translation for 'mfr' Fisher BioReagents missing translation for 'supplierNo' BP29581
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits formation of peptidoglycan polymers of bacterial cell wall

TRUSTED_SUSTAINABILITY

Specifications

Type Vancomycin Hydrochloride
Physical Form Powder/Solid
CAS 1404-93-9
pH 2.5 to 4.5
Source Streptomyces orientalis
Packaging Amber Glass
Potency > 900 U/mg
Water <10.0%
Color White
Quantity 1 g
Molecular Formula C66H76Cl3N9O24
MDL Number MFCD03613611,MFCD03613611
Merck Index 15, 10116
Solubility Information Solubility in water: slight
InChI Key LCTORFDMHNKUSG-UHFFFAOYNA-N
SMILES Cl.CNC(CC(C)C)C(=O)NC1C(O)C2=CC(Cl)=C(OC3=CC4=CC(OC5=C(Cl)C=C(C=C5)C(O)C5NC(=O)C(NC(=O)C4NC(=O)C(CC(N)=O)NC1=O)C1=CC(=C(O)C=C1)C1=C(O)C=C(O)C=C1C(NC5=O)C(O)=O)=C3OC1OC(CO)C(O)C(O)C1OC1CC(C)(N)C(O)C(C)O1)C=C2
IUPAC Name 48-({3-[(4-amino-5-hydroxy-4,6-dimethyloxan-2-yl)oxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl}oxy)-22-(carbamoylmethyl)-5,15-dichloro-2,18,32,35,37-pentahydroxy-19-[4-methyl-2-(methylamino)pentanamido]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentaazaoctacyclo[26.14.2.2³,⁶.2¹⁴,¹⁷.1⁸,¹².1²⁹,³³.0¹⁰,²⁵.0³⁴,³⁹]pentaconta-3,5,8(48),9,11,14,16,29(45),30,32,34,36,38,46,49-pentadecaene-40-carboxylic acid hydrochloride
Molecular Weight (g/mol) 1485.72
PubChem CID 124080918
Formula Weight 1485.73
Chemical Name or Material Vancomycin Hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Vancomycin hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.