missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(V) triisopropoxide oxide, 96%

Product Code. 11453830
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
Unit Size:
100g
1g
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453830 5 g 5g
11443830 1 g 1g
11433830 25 g 25g
11463830 100 g 100g
11423830 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11453830 Supplier Thermo Scientific Alfa Aesar Supplier No. 089798.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5588-84-1 | C9H24O4V | 247.23 g/mol

Vanadium(V) triisopropoxide oxide serves as a building block for a series of neutral oxovanadium complexes containing both ethoxide and acetylacetonate ligands. It is also used in catalysis. Further, it acts as a precursor and used in thermal atomic layer deposition of vanadium pentoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5588-84-1
Molecular Formula C9H24O4V
Molecular Weight (g/mol) 247.23
MDL Number MFCD00015017
InChI Key JOUSPCDMLWUHSO-UHFFFAOYSA-N
Synonym vanadium v oxytriisopropoxide, oxotris propan-2-olato vanadium, vanadium v triisopropoxide oxide, oxotri isopropoxo vanadium, vtip, vo oipr 3, vanadium triisopropoxy oxide, triisopropoxy oxo vanadium, vanadium v tri-isopropoxy oxide, dsggjxauqhkogq-uhfffaoysa-n
PubChem CID 79702
IUPAC Name oxovanadium;propan-2-ol
SMILES O=[V].CC(C)O.CC(C)O.CC(C)O

Specifications

Boiling Point 81°C to 83°C (2mmHg)
Melting Point -12°C
Color Orange-Yellow
Physical Form Liquid
Quantity 5 g
Linear Formula VO[CHO(CH3)2]3
UN Number UN1993
Flash Point 45°C (113°F)
Solubility Information Miscible with toluene,hexane and isopropanol.
Formula Weight 242.2
Percent Purity 96%
Sensitivity Moisture sensitive
Density 1.04 g/mL
Chemical Name or Material Vanadium (V) triisopropoxide oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.