missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(V) trifluoride oxide, 99%, Thermo Scientific™

Product Code. 11367727
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367727 5 g 5g
11377727 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367727 Supplier Thermo Scientific Alfa Aesar Supplier No. 014128.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-31-4
Molecular Formula F3H3OV
Molecular Weight (g/mol) 126.96
MDL Number MFCD00042555
InChI Key HHQFLEDKAVLHOM-UHFFFAOYSA-N
Synonym trifluorooxovanadium, trifluoro oxo vanadium, vanadium oxytrifluoride, vof3, vanadium v trifluoride oxide, vanadium v oxyfluoride, vanadium,trifluorooxo-, t-4-9ci
PubChem CID 123322
IUPAC Name oxovanadium;trihydrofluoride
SMILES F.F.F.O=[V]

Specifications

Melting Point 300°C
Physical Form Powder
Quantity 5 g
UN Number UN3290
Solubility Information Insoluble in water.
Formula Weight 123.94
Percent Purity 99%
Sensitivity Moisture sensitive
Chemical Name or Material Vanadium(V) trifluoride oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.