missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(V) trichloride oxide, V+5 28.5% min

Product Code. 11344919
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344919 100 g 100g
11354919 10 g 10g
11364919 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11344919 Supplier Thermo Scientific Alfa Aesar Supplier No. 081118.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7727-18-6 | Cl3OV | 173.29 g/mol

Vanadium(V) trichloride oxide is used as a catalyst in olefin polymerization reaction, especially for the production of ethylene-propylene rubber. It is also used to synthesis organovanadium compounds and as a mordant in dyeing. It acts as a precursor to prepare vanadium pentoxide, vanadyl dichloride and dioxovanadium monochloride. Further, it serves as a reagent and a strong oxidizing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-18-6
Molecular Formula Cl3OV
Molecular Weight (g/mol) 173.29
MDL Number MFCD00011459
InChI Key WMZFXRASJAIALG-UHFFFAOYSA-K
Synonym vanadyl trichloride, trichlorooxovanadium, vanadium v trichloride oxide, trichlorooxovanadium oxide, vanadium v oxytrichloride, trichlorooxo vanadium, vanadium, trichlorooxo-, t-4, vanadium, trichlorooxo, vanadium oxide trichloride, vanadium trichloride oxide
PubChem CID 24410
IUPAC Name oxovanadium;trihydrochloride
SMILES [Cl-].[Cl-].[Cl-].O=[V+5]

Specifications

Boiling Point 126°C to 127°C
Melting Point -77°C
Color Yellow
Physical Form Liquid
Quantity 100 g
UN Number UN2443
Merck Index 14,9927
Solubility Information Miscible with benzene,dichloromethane and hexane.
Formula Weight 173.3
Concentration or Composition (by Analyte or Components) V+5 28.5% min
Odor Chlorine-like
Sensitivity Moisture sensitive
Density 1.829 g/mL
Chemical Name or Material Vanadium (V) trichloride oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.