missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(V) Oxide, TRC

Product Code. 30005879
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005879 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30005879 Supplier Toronto Research Chemicals Supplier No. TRCV96536010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1314-62-1
Quantity 10 g
Molecular Formula O5 V2
Synonym Vanadium oxide (V2O5), C.I. 77938, Divanadium pentaoxide, Divanadium pentoxide, Pentaoxodivanadium, Red vanadium, VT 1, VT 1 (colorant), Vanadia, Vanadic anhydride, Vanadium oxide, Vanadium oxide (V4O10), Vanadium oxide (VO2.5), Vanadium pentoxide, Vanadium(V) oxide
InChI Formula InChI=1S/5O.2V
SMILES O=[V](=O)O[V](=O)=O
IUPAC Name dioxovanadiooxy(dioxo)vanadium
Molecular Weight (g/mol) 181.88
Formula Weight 181.863
Chemical Name or Material Vanadium(V) oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.