missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(V) oxide, 99.2%, Thermo Scientific Chemicals

Catalog Number 11386108
Click to view available options
Quantity:
25 g
250 g
1 kg
This item is not returnable. View return policy
1314-62-1
O5V2
181.88
MFCD00011457
GNTDGMZSJNCJKK-UHFFFAOYSA-N
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
14814
CHEBI:30045
O=[V](=O)O[V](=O)=O
This item is not returnable. View return policy
1314-62-1
O5V2
181.88
MFCD00011457
GNTDGMZSJNCJKK-UHFFFAOYSA-N
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
14814
CHEBI:30045
O=[V](=O)O[V](=O)=O

As an oxidation catalyst, in refractories, as a ceramics colorant, in the manufacture of vanadium salts, and as a dye.Vanadium pentoxide is the precursor to alloys of vanadium. It is widely used as an industrial catalyst in the manufacturing of sulfuric acid, maleic anhydride, phthalic anhydride, adipic acid, acrylic acid, oxalic acid and anthraquinone. It finds application as a detector material in bolometers and microbolometer arrays for thermal imaging, and as an ethanol sensor in ppm (parts per million) levels. It is also used in vanadium redox batteries, which are used for energy storage in power facilities, such as wind farms. Additionally, it is used as ceramics colorant, as dye and also as a detector material in bolometers in the field of thermal imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • As an oxidation catalyst, in refractories, as a ceramics colorant, in the manufacture of vanadium salts, and as a dye
TRUSTED_SUSTAINABILITY
Boiling Point 1750°C (decomposition)
Melting Point 690°C
Physical Form Powder
Quantity 25 g
UN Number UN2862
Merck Index 14,9921
Solubility Information Slightly soluble in water; Soluble in acids and alkalies.
IUPAC Name dioxovanadiooxy(dioxo)vanadium
Formula Weight 181.88
Percent Purity 99.2%
Odor Odorless
Density 3.357 g/mL
Chemical Name or Material Vanadium(V) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'