Learn More
Vanadium(V) oxide, 99.2%
Description
As an oxidation catalyst, in refractories, as a ceramics colorant, in the manufacture of vanadium salts, and as a dye.Vanadium pentoxide is the precursor to alloys of vanadium. It is widely used as an industrial catalyst in the manufacturing of sulfuric acid, maleic anhydride, phthalic anhydride, adipic acid, acrylic acid, oxalic acid and anthraquinone. It finds application as a detector material in bolometers and microbolometer arrays for thermal imaging, and as an ethanol sensor in ppm (parts per million) levels. It is also used in vanadium redox batteries, which are used for energy storage in power facilities, such as wind farms. Additionally, it is used as ceramics colorant, as dye and also as a detector material in bolometers in the field of thermal imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- As an oxidation catalyst, in refractories, as a ceramics colorant, in the manufacture of vanadium salts, and as a dye
Specifications
Specifications
| Boiling Point | 1750°C (decomposition) |
| Melting Point | 690°C |
| Physical Form | Powder |
| Quantity | 250 g |
| UN Number | UN2862 |
| Merck Index | 14,9921 |
| Solubility Information | Slightly soluble in water; Soluble in acids and alkalies. |
| IUPAC Name | dioxovanadiooxy(dioxo)vanadium |
| Formula Weight | 181.88 |
| Percent Purity | 99.2% |
| Show More |
Safety and Handling
- Vanadium(V) oxide
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 1B
- Reproductive toxicity on or via lactation
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
- H301-Toxic if swallowed.
- H330-Fatal if inhaled.
- H335-May cause respiratory irritation.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H362-May cause harm to breast-fed children.
- H372-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Vanadium oxide
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.