missing translation for 'onlineSavingsMsg'
Learn More

Vanadium standard solution, for AAS, 1mg/mL V(3+) in 2% HNO3, ACROS Organics™

Brand:  Acros Organics 196431000

Code : 29

Additional Details : CAS Number : 7697-37-2 Weight : 0.15000kg Transport : UN number : 3264 Chem class : 1a Pack group : III


Quantity 100mL
Packaging Plastic bottle

 View more versions of this product

Product Code. 10286630

  • £15.00 / 100mL


Estimated Shipment
08-02-2018
Add to basket


Specifications

Chemical Identifiers

Vanadium standard solution
7732-18-5
V
MFCD00011453
1 mg/ml V in 2% HNO3
For AAS
50.94

Specifications

Vanadium standard solution
7732-18-5
Solution
995 to 1005 μg/mL of V(3+) in 2% HNO3 (20°C)
MFCD00011453
Solubility in water: miscible.
100mL
1 mg/ml V in 2% HNO3
For AAS
V
50.94
Plastic bottle
Clear
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Vanadium standard solution, 1 mg/ml V(3+) in 2% HNO3

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P264-Wash face, hands and anyA┬ž thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.

SDS