missing translation for 'onlineSavingsMsg'
Learn More

Vanadium rod, 12.7mm (0.5in) dia, 99.5% (metals basis)

Product Code. p-7048644
Click to view available options
Quantity:
2 cm
10 cm
Unit Size:
10cm
2cm
This item is not returnable. View return policy

Product Code. 11366118

Brand: Thermo Scientific Alfa Aesar 011098.BP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-62-2 | V | 50.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-62-2
Molecular Formula V
Molecular Weight (g/mol) 50.94
MDL Number MFCD00011453
InChI Key LEONUFNNVUYDNQ-UHFFFAOYSA-N
Synonym dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
PubChem CID 23990
ChEBI CHEBI:35170
IUPAC Name vanadium
SMILES [V]

Specifications

Quantity 2 cm
Odor Odorless
Weight ≈7.74g/cm
Form Rod
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Vanadium rod, 12.7mm (0.5 in.) dia.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.