missing translation for 'onlineSavingsMsg'
Learn More

Vanadium, plasma standard solution, Specpure™ V 1000μg/mL

Product Code. p-4408611
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
This item is not returnable. View return policy

Product Code. 11331419

Brand: Thermo Scientific Alfa Aesar 013850.AD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-62-1 | O5V2 | 181.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-62-1, 7697-37-2, 7732-18-5
Molecular Formula O5V2
Molecular Weight (g/mol) 181.88
MDL Number MFCD00011457
InChI Key GNTDGMZSJNCJKK-UHFFFAOYSA-N
PubChem CID 14814
ChEBI CHEBI:30045
IUPAC Name dioxovanadiooxy(dioxo)vanadium
SMILES O=[V](=O)O[V](=O)=O

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Quantity 50 mL
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Vanadium(V) oxide: 0.18%; Water: 94.82%
Chemical Name or Material Vanadium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.