missing translation for 'onlineSavingsMsg'
Learn More

Vanadium pieces, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog Number 11335597
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
7440-62-2
V
50.94
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
23990
CHEBI:35170
vanadium
[V]
This item is not returnable. View return policy
7440-62-2
V
50.94
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
23990
CHEBI:35170
vanadium
[V]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver Gray pieces
Assay Percent Range 99.7% (Metals basis)
Quantity 10 g
Odor Odorless
Form Pieces
Chemical Name or Material Vanadium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'