missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) sulfate oxide hydrate, 99.9% (metals basis)

Product Code. 11306108
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11306108 250 g 250g
11386098 10 g 10g
11396098 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11306108 Supplier Thermo Scientific Alfa Aesar Supplier No. 011090.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123334-20-3 | O5SV | 163.00 g/mol

Vanadium(IV) sulfate oxide hydrate is used as a catalyst for the oxidative aromatization of cyclohexenone. It is also used as a component in some food supplements. It is one of the intermediate during the extraction of vanadium from petroleum residues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123334-20-3
Molecular Formula O5SV
Molecular Weight (g/mol) 163.00
MDL Number MFCD00149933
InChI Key UUUGYDOQQLOJQA-UHFFFAOYSA-L
Synonym Vanadium oxysulfate hydrate
PubChem CID 16211496
SMILES O=[V++].[O-]S([O-])(=O)=O

Specifications

Physical Form Lump
Quantity 250 g
Assay Percent Range 99.9%, metal basis
Solubility Information Soluble in water.
IUPAC Name oxovanadium(2+);sulfate;hydrate
Formula Weight 163.00 (Anhydrous)
Density 2.5
Chemical Name or Material Vanadium(IV) Sulfate Oxide Hydrate
compliance-icons
Product Identifier
  • Vanadium(IV) sulfate oxide hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.