missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) sulfate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11396098
Click to view available options
Quantity:
10 g
50 g
250 g
123334-20-3
O5SV
163.00
MFCD00149933
UUUGYDOQQLOJQA-UHFFFAOYSA-L
Vanadium oxysulfate hydrate
16211496
O=[V++].[O-]S([O-])(=O)=O
123334-20-3
O5SV
163.00
MFCD00149933
UUUGYDOQQLOJQA-UHFFFAOYSA-L
Vanadium oxysulfate hydrate
16211496
O=[V++].[O-]S([O-])(=O)=O

Vanadium(IV) sulfate oxide hydrate is used as a catalyst for the oxidative aromatization of cyclohexenone. It is also used as a component in some food supplements. It is one of the intermediate during the extraction of vanadium from petroleum residues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Lump
Quantity 50 g
Assay Percent Range 99.9%, metal basis
Solubility Information Soluble in water.
IUPAC Name oxovanadium(2+);sulfate;hydrate
Formula Weight 163.00 (Anhydrous)
Density 2.5
Chemical Name or Material Vanadium(IV) Sulfate Oxide Hydrate
compliance-icons
Product Identifier
  • Vanadium(IV) sulfate oxide hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'