missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium(IV) oxide phthalocyanine, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
2 g
10 g
Description
It is used as an organic light-emitting diode intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Blue to Purple |
Physical Form | Crystalline |
Quantity | 0.5 g |
UN Number | UN3285 |
Solubility Information | Insoluble in water. |
Formula Weight | 579.45 |
Chemical Name or Material | Vanadium(IV) oxide phthalocyanine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'