missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) oxide meso-tetraphenylporphine, Thermo Scientific Chemicals

Product Code. 11460231
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460231 100 mg 100mg
11314497 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460231 Supplier Thermo Scientific Alfa Aesar Supplier No. 039583.MC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is used for research in catalysis. Thin films of VOTPP may be used in detecting methane gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14705-63-6
Molecular Formula C44H30N4O2V
Molecular Weight (g/mol) 697.692
MDL Number MFCD00012154
InChI Key LRQXAIDMLNPKOR-UHFFFAOYSA-N
Synonym 5,10,15,20-tetraphenyl-21h,23h-porphine vanadium iv oxide
PubChem CID 131699123
IUPAC Name oxygen(2-);5,10,15,20-tetraphenyl-21,22-dihydroporphyrin;vanadium(4+)
SMILES C1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)N3.[O-2].[O-2].[V+4]

Specifications

Color Purple
Physical Form Crystalline
Quantity 100 mg
Solubility Information Insoluble in water.
Formula Weight 679.67
Odor Odorless
Chemical Name or Material Vanadium(IV) oxide meso-tetraphenylporphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.