missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) oxide bis(2,4-pentanedionate)

Product Code. 11394919
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394919 250 g 250g
11374919 10 g 10g
11384919 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394919 Supplier Thermo Scientific Alfa Aesar Supplier No. 081119.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment | CAS: 3153-26-2 | C10H14O5V | 265.16 g/mol

Employed as a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment. It is used in organic chemistry as a reagent in the epoxidation of allylic alcohols in combination with tert-butyl hydroperoxide (TBHP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3153-26-2
Molecular Formula C10H14O5V
Molecular Weight (g/mol) 265.16
MDL Number MFCD00000032
InChI Key JFHJZWAQYMGNBE-SUKNRPLKSA-L
Synonym vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide
PubChem CID 131674261
IUPAC Name (Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate
SMILES O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Boiling Point 140°C (13mmHg)
Melting Point 256°C to 259°C
Physical Form Powder
Quantity 250 g
UN Number UN3285
Solubility Information Moderately soluble in acetone, ether and chloroform; Soluble in ethanol and benzene
Formula Weight 265.16
Density 1.4 g/mL
Chemical Name or Material Vanadium (IV) oxide bis (2,4-pentanedionate)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.