missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) Oxide Acetylacetonate, TRC

Product Code. 30449561
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449561 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449561 Supplier Toronto Research Chemicals Supplier No. TRCV09636010G
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vanadium(IV) Oxide Acetylacetonate
CAS 3153-26-2
Molecular Formula C10H14O5V
Quantity 10 g
Synonym Bis(2,4-pentanedionato)oxovanadium, Bis(2,4-pentanedionato)oxovanadium(IV), Bis(acetylacetonato)oxovanadium, Bis(acetylacetonato)oxovanadium(IV), Bis(acetylacetonato)oxyvanadium, NSC 116105, NSC 4659, NSC 52327, Nacem Vanadyl, Oxobis(2,4-pentanedionato)vanadium, Oxobis(acetylacetonato)vanadium, Oxovanadium(II) Acetylacetonate, Vanadium Oxide Acetylacetonate, Vanadium(IV) Bis(acetylacetonato)(oxo), Vanadium(IV) Oxide Bis(2,4-pentanedionate), Vanadyl Acetylacetonate, Vanadyl Bis(2,4-pentanedionate), Vanadyl Bis(acetylacetonate), VoAA
InChI Formula InChI=1S/2C5H8O2.O.V/c2*1-4(6)3-5(2)7;;/h2*3,6H,1-2H3;;/q;;;+2/p-2/b2*4-3-;;
SMILES CC(=O)\C=C(\C)/O[V](=O)O\C(=C/C(=O)C)\C
IUPAC Name oxo-bis[[(Z)-4-oxopent-2-en-2-yl]oxy]vanadium
Molecular Weight (g/mol) 265.16
Formula Weight 265.0281
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.