missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) oxide, 99% (metals basis)

Product Code. 11398227
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398227 50 g 50g
11308237 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398227 Supplier Thermo Scientific Alfa Aesar Supplier No. 022957.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12036-21-4 | O2V | 82.94 g/mol

Vanadium(IV) oxide is used in chemical sensors, energy-conserving coatings, transparent conductors and switching materials. It acts as a stationary optical shutter, optical modulators, cameras and data storage. As infrared modulators, it is used for the missile guidance systems. Thin films made up of vanadium dioxide find applications in electro-optical switches, micro-optical switch, passive smart radiators and sunshields for spacecraft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-21-4
Molecular Formula O2V
Molecular Weight (g/mol) 82.94
MDL Number MFCD00049702
InChI Key GRUMUEUJTSXQOI-UHFFFAOYSA-N
Synonym vanadium dioxide, vanadium iv oxide, vanadium oxide vo2, vo2, vanadium iv oxide trace metals basis 10g
PubChem CID 82849
ChEBI CHEBI:30047
IUPAC Name dioxovanadium
SMILES O=[V]=O

Specifications

Melting Point 1967°C
Physical Form Powder
Quantity 50 g
Assay Percent Range 99% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 82.94
Density 4.339
Chemical Name or Material Vanadium(IV) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.