missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) fluoride, 95%

Product Code. 11327247
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327247 5 g 5g
11317247 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327247 Supplier Thermo Scientific Alfa Aesar Supplier No. 011543.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10049-16-8

Vanadium(IV) fluoride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-16-8
MDL Number MFCD00042554
Synonym vanadium iv fluoride, acmc-20aja2, vanadium 4+ tetrafluoride, vanadium 4+ ion tetrafluoride, fluoride, fluoride, fluoride, vanadium salt, fluoride

Specifications

Melting Point 325°C (decomposition)
Color Yellow to Brown
Physical Form Powder
Quantity 5 g
Linear Formula VF4
UN Number UN2923
Solubility Information Soluble in water (blue color), acetone (deep green color), and glacial acetic acid (blue-green color)
Formula Weight 126.94
Percent Purity 95%
Sensitivity Moisture sensitive
Density 2.975 g/mL
Chemical Name or Material Vanadium(IV) fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.