missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) chloride, 99+%

Product Code. 10165840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10165840 100 g Glass bottle 100g
10742301 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10165840 Supplier Thermo Scientific Acros Supplier No. 299691000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7632-51-1 | Cl4V | 192.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7632-51-1
Molecular Formula Cl4V
Molecular Weight (g/mol) 192.75
MDL Number MFCD00061343
InChI Key JTJFQBNJBPPZRI-UHFFFAOYSA-J
Synonym vanadium tetrachloride, vanadium iv chloride, vcl4, vanadium chloride vcl4 , t-4, vanadium chloride vcl4, vanadium chloride, t-4, vanadium tetrachloride un2444 corrosive
PubChem CID 24273
IUPAC Name tetrachlorovanadium
SMILES Cl[V](Cl)(Cl)Cl

Specifications

Boiling Point 153.0°C
Melting Point -20.5°C
Color Brown-Red
Physical Form Liquid
Quantity 100 g
Assay Percent Range 99% min. (VCl4)
Linear Formula VCl4
Solubility Information Solubility in water: vigorous reaction. Other solubilities: soluble in absolute alcohol, ether, chloroform,, acetic acid
Formula Weight 192.75
Percent Purity ≥99%
Packaging Glass bottle
Specific Gravity 1.82
Density 1.8200g/mL
Chemical Name or Material Vanadium(IV) chloride, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.