missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) chloride, 99% (metals basis)

Product Code. 11311579
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11311579

Brand: Thermo Scientific Alfa Aesar 014039.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7718-98-1 | Cl3V | 157.29 g/mol

Vanadium(III) chloride acts as an intermediate in organic synthesis and in the manufacture of organic light emitting diodes. It acts as a precursor to other vanadium(III) complexes. Further, it is used in the preparation of vanadium dichloride and organovanadium compounds. In addition to this, it serves as a catalyst in the production of ethylene propylene diene monomer rubber and polyethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7718-98-1
Molecular Formula Cl3V
Molecular Weight (g/mol) 157.29
MDL Number MFCD00011454
InChI Key HQYCOEXWFMFWLR-UHFFFAOYSA-K
PubChem CID 62647
IUPAC Name trichlorovanadium
SMILES [Cl-].[Cl-].[Cl-].[V+3]

Specifications

Quantity 50 g
Assay Percent Range 99% (metals basis)
Solubility Information Soluble in tetrahydrofuran,alcohol,ether and hydrocarbons.
Formula Weight 157.3
Sensitivity Light and moisture sensitive
Density 3
Chemical Name or Material Vanadium(III) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.