missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) bromide, 99.5% (metals basis)

Product Code. 11344929
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344929 10 g 10g
11334929 2 g 2g
11324929 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11344929 Supplier Thermo Scientific Alfa Aesar Supplier No. 081135.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13470-26-3 | Br3V | 290.654 g/mol

Vanadium(III) bromide is used as a catalyst for C(sp3)-H fluorination reactions especially the benzylic fluorination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13470-26-3
Molecular Formula Br3V
Molecular Weight (g/mol) 290.654
MDL Number MFCD00049695
InChI Key ZOYIPGHJSALYPY-UHFFFAOYSA-K
Synonym vanadium bromide, vanadium tribromide, vanadium bromide vbr3, vanadium iii bromide, wln: va e3, vanadium iii bromide-v
PubChem CID 83509
IUPAC Name tribromovanadium
SMILES [V](Br)(Br)Br

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 10 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Soluble in water and tetrahydrofuran.
Formula Weight 290.67
Sensitivity Moisture sensitive
Density 4.00^1^8
Chemical Name or Material Vanadium(III) bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.