missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAMP8 Recombinant Rabbit Monoclonal Antibody (5H9S4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542860
This item is not returnable.
View return policy
Description
Positive test controls include: Mouse liver, Mouse kidney, HeLa, K-562, Rat lung. Immunogen sequence: MEEASEGGGN DRVRNLQSEV EGVKNIMTQN VERILARGEN LEHLRNKTED LEATSEHFKT TSQKVARKFW WKNVKMIVLI CVIVFIIILF IVLFATGAFS.
Involved in the targeting and/or fusion of transport vesicles to their target membrane. Involved for dense-granule secretion in platelets. Plays a role in regulated enzyme secretion in pancreatic acinar cells. Involved in the abscission of the midbody during cell division, which leads to completely separate daughter cells. Involved in the homotypic fusion of early and late endosomes.
Specifications
| VAMP8 | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70404, Q9BV40, Q9WUF4 | |
| VAMP8 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human VAMP8 (NP_0037522). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 5H9S4 | |
| Unconjugated | |
| VAMP8 | |
| AU041171; Edb; endobrevin; hypothetical protein LOC507309; vamp; VAMP8; VAMP-8; vesicle associated membrane protein 8; vesicle-associated membrane protein 8; vesicle-associated membrane protein 8 (endobrevin); wu:fa18d07; zgc:56588 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22320, 83730, 8673 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction