missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAMP8 Monoclonal Antibody (1414CT354.12.23.93)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537661
This item is not returnable.
View return policy
Description
VAMP8 Monoclonal Antibody for Western Blot, IHC (P)
Involved in the targeting and/or fusion of transport vesicles to their target membrane. Involved for dense-granule secretion in platelets. Plays a role in regulated enzyme secretion in pancreatic acinar cells. Involved in the abscission of the midbody during cell division, which leads to completely separate daughter cells. Involved in the homotypic fusion of early and late endosomes.
Specifications
| VAMP8 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9BV40 | |
| VAMP8 | |
| KLH conjugated synthetic peptide between 2~24 amino acids from the N-terminal region of human VAMP8. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1414CT354.12.23.93 | |
| Unconjugated | |
| VAMP8 | |
| AU041171; Edb; endobrevin; hypothetical protein LOC507309; vamp; VAMP8; VAMP-8; vesicle associated membrane protein 8; vesicle-associated membrane protein 8; vesicle-associated membrane protein 8 (endobrevin); wu:fa18d07; zgc:56588 | |
| Mouse | |
| Protein G | |
| RUO | |
| 8673 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction