missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP7/TI-VAMP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22268-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to VAMP7.
This gene encodes a transmembrane protein that is a member of the soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) family. The encoded protein localizes to late endosomes and lysosomes and is involved in the fusion of transport vesicles to their target membranes. Alternate splicing results in multiple transcript variants.Specifications
| VAMP7/TI-VAMP | |
| Polyclonal | |
| Unconjugated | |
| VAMP7 | |
| SYBL1, Synaptobrevin like protein 1, Tetanus insensitive VAMP, TI VAMP, TI-VAMP, VAMP 7, VAMP7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20955, 6845, 85491 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51809, P70280, Q9JHW5 | |
| Vamp7 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction