missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAMP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583348
This item is not returnable.
View return policy
Description
Immunogen sequence: YAMNSEFSSV LAAQLKHHSE NKGLDKVMET QAQVDELKGI MVRNIDLVAQ RGERLELLID KTENLVDSSV TFKTTSRNLA RAMCMKNL Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 98%.
This gene encodes a transmembrane protein that is a member of the soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) family. The encoded protein localizes to late endosomes and lysosomes and is involved in the fusion of transport vesicles to their target membranes. Alternate splicing results in multiple transcript variants.
Specifications
| VAMP7 | |
| Polyclonal | |
| Unconjugated | |
| Vamp7 | |
| Sybl1; synaptobrevin-like 1; synaptobrevin-like protein 1; tetanus neurotoxin-insensitive VAMP; tetanus-insensitive VAMP; TIVAMP; Ti-VAMP; vamp; Vamp7; VAMP-7; vesicle associated membrane protein 7; vesicle-associated membrane protein 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6845 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51809 | |
| Vamp7 | |
| Recombinant protein corresponding to Human VAMP7. Recombinant protein control fragment (Product #RP-96560). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction