missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAMP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534405
This item is not returnable.
View return policy
Description
A suggested positive control is mouse lung tissue lysate. PA5-34405 can be used with blocking peptide PEP-1447.
This gene encodes a transmembrane protein that is a member of the soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) family. The encoded protein localizes to late endosomes and lysosomes and is involved in the fusion of transport vesicles to their target membranes. Alternate splicing results in multiple transcript variants.
Specifications
| VAMP7 | |
| Polyclonal | |
| Unconjugated | |
| Vamp7 | |
| Sybl1; synaptobrevin-like 1; synaptobrevin-like protein 1; tetanus neurotoxin-insensitive VAMP; tetanus-insensitive VAMP; TIVAMP; Ti-VAMP; vamp; Vamp7; VAMP-7; vesicle associated membrane protein 7; vesicle-associated membrane protein 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20955, 6845, 85491 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P51809, P70280, Q9JHW5 | |
| Vamp7 | |
| An 18 amino acid synthetic peptide near the amino terminus of human VAMP7. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction