missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAMP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525755
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine based on sequence homology.
The vesicle associated membrane proteins (VAMP) or synaptobrevins are calcium binding proteins specific to eukaryotes. VAMPs, along with synaptosomal associated protein of 25 kDa (SNAP-25) and syntaxin, form the core complex of soluble NSF attachment protein receptor (SNARE) proteins that interact with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and alpha-SNAP. These membrane associated proteins play a key role in the regulation of vesicle membrane fusion with the plasma membrane. The Clostridium tetani neurotoxin is a metalloprotease with specificity for VAMP. In Alzheimer's disease, VAMP levels of all isoforms appear to be significantly lowered.
Specifications
| VAMP4 | |
| Polyclonal | |
| Unconjugated | |
| VAMP4 | |
| D1Ertd147e; vamp; VAMP24; VAMP4; VAMP-4; vesicle associated membrane protein 4; vesicle-associated membrane protein 4 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 53330, 8674 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.45 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O70480, O75379 | |
| VAMP4 | |
| KLH conjugated synthetic peptide between 16-45 amino acids from the central region of human VAMP4. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction