missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP3/Cellubrevin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | VAMP3/Cellubrevin |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228328
|
Novus Biologicals
NBP3-38430-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228262
|
Novus Biologicals
NBP3-38430-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VAMP3/Cellubrevin Polyclonal antibody specifically detects VAMP3/Cellubrevin in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| VAMP3/Cellubrevin | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neurotransmission | |
| PBS (pH 7.3), 50% glycerol | |
| 9341 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CEBCellubrevin, cellubrevin, SYB3, Synaptobrevin-3, VAMP-3, vesicle-associated membrane protein 3, vesicle-associated membrane protein 3 (cellubrevin) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human VAMP3/Cellubrevin (NP_004772.1).,, Sequence:, MSTGPTAATGSNRRLQQTQNQVDEVVDIMRVNVDKVLERDQKLSELDDRADALQAGASQFETSAAKLKRKYWWKNCKMWA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title