missing translation for 'onlineSavingsMsg'
Learn More

VAMP3/Cellubrevin Antibody (4G9), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18405083 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405083 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405083 Supplier Novus Biologicals Supplier No. NBP197948MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VAMP3/Cellubrevin Monoclonal antibody specifically detects VAMP3/Cellubrevin in Human, Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAMP3/Cellubrevin
Applications Flow Cytometry
Classification Monoclonal
Clone 4G9
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CEBCellubrevin, cellubrevin, SYB3, Synaptobrevin-3, VAMP-3, vesicle-associated membrane protein 3, vesicle-associated membrane protein 3 (cellubrevin)
Host Species Mouse
Immunogen Recombinant human VAMP3/Cellubrevin (1-77aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 9341
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.