Learn More
Invitrogen™ VAMP1/VAMP2/VAMP3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5103178
Product Code. 16619634
  • £448.00 / 100µL
Description
Antibody detects endogenous levels of total VAMP 1/2/3.
Synapotobrevins, syntaxins, and the synaptosomal-associated protein SNAP25 are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. The protein encoded by this gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Mutations in this gene are associated with autosomal dominant spastic ataxia 1. Multiple alternative splice variants have been described, but the full-length nature of some variants has not been defined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VAMP1/VAMP2/VAMP3
Polyclonal
Unconjugated
VAMP1
CEB; Cellubrevin; D130027G05Rik; lew; RATVAMPB; RATVAMPIR; SPAX1; SYB; SYB1; Syb-1; SYB2; Syb-2; SYB3; sybII; synaptobrevin 1; synaptobrevin 2; Synaptobrevin 2 (vesicle-associated membrane protein VAMP-2); synaptobrevin II; synaptobrevin-1; Synaptobrevin-2; synaptobrevin-3; VAMP1; VAMP-1; VAMP2; VAMP-2; Vamp3; VAMP-3; vesicle associated membrane protein 1; vesicle associated membrane protein 2; vesicle associated membrane protein 3; vesicle-associated membrane 2; Vesicle-associated membrane protein (synaptobrevin 2); vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1); vesicle-associated membrane protein 2; vesicle-associated membrane protein 2 (synaptobrevin 2); vesicle-associated membrane protein 3
Rabbit
Affinity chromatography
RUO
22317, 22318, 22319, 24803, 25624, 29528, 6843, 6844, 9341
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P23763, P63024, P63025, P63027, P63044, P63045, Q15836, Q62442, Q63666
VAMP1, Vamp2, Vamp3
A synthesized peptide derived from human VAMP 1/2/3, corresponding to a region within C-terminal amino acids.
100 μL
Primary
Human, Mouse, Rat