missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP-7 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41183H
This item is not returnable.
View return policy
Description
VAMP-7 Polyclonal antibody specifically detects VAMP-7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| VAMP-7 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| HRP | |
| FLJ54296, SYBL1FLJ53762, synaptobrevin-like 1, Synaptobrevin-like protein 1, tetanus neurotoxin-insensitive VAMP, Tetanus-insensitive VAMP, Ti-VAMP, TIVAMP, VAMP-7FLJ53045, vesicle-associated membrane protein 7 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human VAMP7. The immunogen is located within amino acids 20 - 70 of VAMP7. Amino Acid Squence: AKIPSENNKLTYSHGNYL | |
| 0.1 mL | |
| Endosome Markers | |
| 6845 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction