missing translation for 'onlineSavingsMsg'
Learn More

VAMP-5 Antibody (3E5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£169.00 - £319.00

Specifications

Antigen VAMP-5
Clone 3E5
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Applications Western Blot, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18652847
View Documents
Novus Biologicals
NBP2-50357-0.025ml
0.025 mL
£169.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18677547
View Documents
Novus Biologicals
NBP2-50357
0.1 mL
£319.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VAMP-5 Monoclonal antibody specifically detects VAMP-5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

VAMP-5
1 mg/mL
Western Blot, Immunofluorescence
Unconjugated
Mouse
Membrane Vesicle Markers, Neuronal Cell Markers, Neurotransmission
PBS
10791
IgG2b
Protein A purified
3E5
Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Monoclonal
Purified
RUO
Human
myobrevin, VAMP-5, vesicle-associated membrane protein 5, vesicle-associated membrane protein 5 (myobrevin)
GST-mVAMP-5-II and KLH-mVAMP-5-II peptide sequences
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.