missing translation for 'onlineSavingsMsg'
Learn More

VAMP-2 Antibody [DyLight 405], Novus Biologicals™

Product Code. 30737832 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737832 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737832 Supplier Novus Biologicals Supplier No. AF5136E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

VAMP-2 Polyclonal antibody specifically detects VAMP-2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAMP-2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias FLJ11460, SYB2vesicle-associated membrane protein 2, synaptobrevin 2, Synaptobrevin-2, VAMP-2vesicle associated membrane protein 2, vesicle-associated membrane protein 2 (synaptobrevin 2)
Host Species Goat
Immunogen E. coli-derived recombinant human VAMP-2, Met1-Lys94, Accession # P63027
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6844
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.