missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP-1 Rabbit anti-Human, Mouse, Rat, Clone: 1A3K6, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | VAMP-1 |
|---|---|
| Clone | 1A3K6 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18342116
|
Novus Biologicals
NBP3-16212-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18340326
|
Novus Biologicals
NBP3-16212-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VAMP-1 Monoclonal antibody specifically detects VAMP-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| VAMP-1 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp686H12131, SYB1vesicle-associated membrane protein 1, synaptobrevin 1, Synaptobrevin-1, VAMP-1synaptobrevin-1, vesicle-associated membrane protein 1 (synaptobrevin 1) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-118 of human VAMP-1 (P23763). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 1A3K6 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Membrane Trafficking and Chaperones, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6843 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title