missing translation for 'onlineSavingsMsg'
Learn More

SCAT Valve Coupling Units

Product Code. 16615002 Shop All SCAT Europe Products
Change view
Click to view available options
Components: Coupling (m)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16615002 Components: Coupling (m)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16615002 Supplier SCAT Supplier No. 160193

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quick disconnect couplings allow safe and rapid connection of tubing.

  • Disconnection with a single click
  • Integrated valve function: closed safety system is maintained even when a tube is disconnected
  • Practical quick disconnect couplings can be used for all SCAT safety systems

Specifications

Description Valved In-Line Coupling Body (f), PP, for tubes approx 6 mm ID with screw connection, Seal of EPDM
Model Valved In-Line Coupling Body (f)
Quantity 1 Component
For Use With (Equipment) 6 mm Tube with Screw Conncection
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.