Learn More
Cytiva (formerly Pall Lab) ValuPrep™ Syringe Filters with Nylon Membrane
Syringe filters perform sample clean-up and preparation.
Brand: Cytiva (formerly Pall Lab) 4118
Additional Details : Weight : 3.15400kg
Description
- Ensures the purest samples are analyzed in liquid chromatography, mass spectrometry, and ion chromatography applications
Specifications
Nylon | |
Non-sterile | |
1000/Pk. |
0.45 μm | |
2.8 cm2 | |
25 mm |