missing translation for 'onlineSavingsMsg'
Learn More

Valsartan, MedChemExpress

Product Code. 16477478
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16477478

Brand: Medchem Express HY18204

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Valsartan (CGP 48933) is an angiotensin II receptor antagonist and has the potential for high blood pressure and heart failure research.

Valsartan (CGP 48933) is an angiotensin II receptor antagonist and has the potential for high blood pressure and heart failure research.

Specifications

Chemical Name or Material Valsartan
Physical Form Solid
CAS 137862-53-4
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H29N5O3
For Use With (Application) COVID-19-immunoregulation
Synonym CGP 48933
Solubility Information DMSO : ≥ 100 mg/mL (229.61 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CC(C)[C@@H](C(O)=O)N(C(CCCC)=O)CC1=CC=C(C2=CC=CC=C2C3=NNN=N3)C=C1
Molecular Weight (g/mol) 435.52
Formula Weight 435.52
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.