missing translation for 'onlineSavingsMsg'
Learn More

Valsartan D8 (valine D8), Dr. Ehrenstorfer

Product Code. 30321020
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321020 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321020 Supplier Dr. Ehrenstorfer Supplier No. DREC17899993
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-(-)-Valsartan-d8 (valine-d8)
CAS 1089736-72-0
Molecular Formula C24H21D8N5O3
Quantity 10 mg
Synonym N-(1-Oxopentyl)-N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-L-valine-2,3,4,4,4,4',4',4'-d8;
InChI Formula InChI=1S/C24H29N5O3/c1-4-5-10-21(30)29(22(16(2)3)24(31)32)15-17-11-13-18(14-12-17)19-8-6-7-9-20(19)23-25-27-28-26-23/h6-9,11-14,16,22H,4-5,10,15H2,1-3H3,(H,31,32)(H,25,26,27,28)/t22-/m0/s1/i2D3,3D3,16D,22D
SMILES [2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])[C@]([2H])(N(Cc1ccc(cc1)c2ccccc2c3nnn[nH]3)C(=O)CCCC)C(=O)O
IUPAC Name (2S)-2,3,4,4,4-pentadeuterio-2-[pentanoyl-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]-3-(trideuteriomethyl)butanoic acid
Molecular Weight (g/mol) 443.58
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.