missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ValRS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15931-1-AP-150UL
This item is not returnable.
View return policy
Description
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. The protein encoded by this gene belongs to class-I aminoacyl-tRNA synthetase family and is located in the class III region of the major histocompatibility complex.Specifications
| ValRS | |
| Polyclonal | |
| Unconjugated | |
| VARS | |
| G7A, Protein G7a, Valine tRNA ligase, ValRS, Valyl-tRNA synthetase, VARS, VARS1, VARS2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22321, 25009, 7407 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P26640, Q04462, Q9Z1Q9 | |
| VARS | |
| ValRS Fusion Protein Ag8778 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction