missing translation for 'onlineSavingsMsg'
Learn More

Valpromide, MedChemExpress

Product Code. 16444647
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444647 50 mg 50mg
16438738 10 mg 10mg
16418846 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16444647 Supplier Medchem Express Supplier No. HYB2117
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Valpromide is an amide derivative of valproic acid and inhibits human epoxide hydrolase.

Valpromide is an amide derivative of valproic acid and inhibits human epoxide hydrolase.

Specifications

Chemical Name or Material Valpromide
CAS 2430-27-5
Color White
Purity Grade Notes Research
Molecular Formula C8H17NO
Quantity 50 mg
Solubility Information DMSO : ≥ 50 mg/mL (349.09 mM)
SMILES CCCC(CCC)C(N)=O
Molecular Weight (g/mol) 143.23
Formula Weight 143.23
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.