missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valproic Acid Semisodium Salt, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Valproic Acid Semisodium Salt |
| CAS | 76584-70-8 |
| Molecular Formula | C8 H16 O2 . C8 H15 O2 . Na |
| Quantity | 500 mg |
| Synonym | Abbott 50711, Depakote, Divalproex sodium, Epival, Sodium hydrogen bis(2-propylpentanoate), Sodium hydrogen bis(2-propylvalerate), Sodium hydrogen divalproate, Valdisoval, Valproate semisodium, Pentanoic acid','2-propyl-','sodium salt (2:1) |
| InChI Formula | InChI=1S/2C8H16O2.Na/c2*1-3-5-7(6-4-2)8(9)10;/h2*7H,3-6H2,1-2H3,(H,9,10);/q;;+1/p-1 |
| SMILES | [Na+].CCCC(CCC)C(=O)O.CCCC(CCC)C(=O)[O-] |
| IUPAC Name | sodium;2-propylpentanoate;2-propylpentanoic acid |
| Molecular Weight (g/mol) | 310.4 |
| Formula Weight | 310.212 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction