missing translation for 'onlineSavingsMsg'
Learn More

Valproic Acid Semisodium Salt, TRC

Product Code. 30449253
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30449253

Brand: Toronto Research Chemicals TRCV0947555G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Valproic Acid Semisodium Salt
CAS 76584-70-8
Molecular Formula C8 H16 O2 . C8 H15 O2 . Na
Quantity 5 g
Synonym Abbott 50711, Depakote, Divalproex sodium, Epival, Sodium hydrogen bis(2-propylpentanoate), Sodium hydrogen bis(2-propylvalerate), Sodium hydrogen divalproate, Valdisoval, Valproate semisodium, Pentanoic acid','2-propyl-','sodium salt (2:1)
InChI Formula InChI=1S/2C8H16O2.Na/c2*1-3-5-7(6-4-2)8(9)10;/h2*7H,3-6H2,1-2H3,(H,9,10);/q;;+1/p-1
SMILES [Na+].CCCC(CCC)C(=O)O.CCCC(CCC)C(=O)[O-]
IUPAC Name sodium;2-propylpentanoate;2-propylpentanoic acid
Molecular Weight (g/mol) 310.4
Formula Weight 310.212
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.