Learn More
Invitrogen™ Valproic Acid Monoclonal Antibody (VP10-201.7), Invitrogen™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA110766
 View more versions of this product
Product Code. 11519032
  • £478.00 / 500µg
Description
MA1-10766 detects Valproic Acid. MA1-10766 has been successfully used in ELISA applications. The MA1-10766 immunogen is Valproic acid, conjugated to KLH.
Valproic acid, valproate sodium, and divalproex belong to the group of medicines called anticonvulsants. They are used to control certain types of seizures in the treatment of epilepsy. Also Valproic acid relieves histone deacetylases (HDACs) dependent transcriptional repression and causes hyperacetylation of histones in cultured cells and in vivo. Valproic acid inhibits HDAC activity in vitro, most probably by binding to the catalytic center of HDACs. Most importantly, valproic acid induces differentiation of carcinoma cells, transformed hematopoietic progenitor cells and leukemic blasts from acute myeloid leukemia patients. More over, tumor growth and metastasis formation are significantly reduced in animal experiments. Therefore, valproic acid might serve as an effective drug for cancer therapy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Valproic Acid
Monoclonal
1.5 mg/mL
PBS with no preservative; pH 7.4
Mouse
Protein A
RUO
Chemical
Antibody
IgG1
ELISA
VP10-201.7
Unconjugated
C8H16O2; Depakene; Depakine; Dipropylacetic acid
Valproic acid, conjugated to KLH.
500 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical