missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Valproic Acid Monoclonal Antibody (VP10-115.2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA5-46132
This item is not returnable.
View return policy
Description
MA1-10765 detects Valproic Acid. MA1-10765 has been successfully used in ELISA applications. The MA1-10765 immunogen is Valproic acid, conjugated to KLH.
Valproic acid, valproate sodium, and divalproex belong to the group of medicines called anticonvulsants. They are used to control certain types of seizures in the treatment of epilepsy. Also Valproic acid relieves histone deacetylases (HDACs) dependent transcriptional repression and causes hyperacetylation of histones in cultured cells and in vivo. Valproic acid inhibits HDAC activity in vitro, most probably by binding to the catalytic center of HDACs. Most importantly, valproic acid induces differentiation of carcinoma cells, transformed hematopoietic progenitor cells and leukemic blasts from acute myeloid leukemia patients. More over, tumor growth and metastasis formation are significantly reduced in animal experiments. Therefore, valproic acid might serve as an effective drug for cancer therapy.
Specifications
| Valproic Acid | |
| Monoclonal | |
| 1.3 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
| ELISA | |
| VP10-115.2 | |
| Unconjugated | |
| C8H16O2; Depakene; Depakine; Dipropylacetic acid | |
| Valproic acid, conjugated to KLH. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction