missing translation for 'onlineSavingsMsg'
Learn More

Valproic Acid EP Impurity E, Mikromol™

Product Code. 30459319
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459319 4 x 25 mg 100mg
17912396 25 mg 25mg
17990436 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30459319 Supplier Mikromol Supplier No. MM0751.0500254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Valproic Acid EP Impurity E, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pentanamide (Valeramide)
CAS 626-97-1
Analyte or Component Names Pentanamide (Valeramide)
Shipping Condition Room Temperature
Molecular Formula C5 H11 N O
Certifications/Compliance ISO 9001
Synonym Pentanamide, Valproic Acid Imp. E (EP), Valeramide, Valproic Acid Impurity E
InChI Formula InChI=1S/C5H11NO/c1-2-3-4-5(6)7/h2-4H2,1H3,(H2,6,7)
Solution Type Neat
SMILES CCCCC(=O)N
IUPAC Name pentanamide
Molecular Weight (g/mol) 101.15
Quantity 4 x 25 mg
Formula Weight 101.0841 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.